missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/CD90.1 Mouse, anti-Rat, Alexa Fluor 488, Clone: OX-7, STEMCELL Technologies
Mouse Monoclonal Antibody
POA - £414.45
Specifications
| Antigen | CD90/CD90.1 |
|---|---|
| Clone | OX-7 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17168561
|
Stemcell Technologies
60024AD.1 |
25 μg | |||||||
17158561
|
Stemcell Technologies
60024AD |
100 μg |
£414.45
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse CD90.1 (Thy-1.1), the latter being an allelic form of CD90 expressed by mouse strains AKR/J, PL, and FVB/N
- The OX-7 antibody does not react with CD90.2, which is expressed by many mouse strains, including CBA and BALB/c
- CD90 is a GPI-linked membrane glycoprotein and member of the immunoglobulin superfamily
- The 25 kDa core protein is N-glycosylated at three sites, giving rise to molecules with a range of molecular masses (25 - 37 kDa)
- In the rat, CD90 is expressed by several cell types, including hematopoietic stem cells, immature B cells, thymocytes and neurons
- This antibody clone has been verified for purity assessments of cells isolated from compatible mouse strains with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851)
Specifications
| CD90/CD90.1 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| Guinea Pig, Mouse, Rabbit, Rat | |
| Primary |
| OX-7 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title