Learn More
CD90/CD90.1 Mouse, anti-Rat, Alexa Fluor 488, Clone: OX-7, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60024AD.1
This item is not returnable.
View return policy
Description
- The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse CD90.1 (Thy-1.1), the latter being an allelic form of CD90 expressed by mouse strains AKR/J, PL, and FVB/N
- The OX-7 antibody does not react with CD90.2, which is expressed by many mouse strains, including CBA and BALB/c
- CD90 is a GPI-linked membrane glycoprotein and member of the immunoglobulin superfamily
- The 25 kDa core protein is N-glycosylated at three sites, giving rise to molecules with a range of molecular masses (25 - 37 kDa)
- In the rat, CD90 is expressed by several cell types, including hematopoietic stem cells, immature B cells, thymocytes and neurons
- This antibody clone has been verified for purity assessments of cells isolated from compatible mouse strains with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851)
Specifications
| CD90/CD90.1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 25 μg | |
| Guinea Pig, Mouse, Rabbit, Rat | |
| T Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OX-7 | |
| Mouse | |
| Primary | |
| IgG1 κ |