missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD9 Immunobeads for Exosome Isolation
Shop All Bio Techne Products
Click to view available options
Quantity:
10 Reactions
Unit Size:
1 set
Specifications
Specifications
| Synonym | EV isolation, exosome isolation, extracellular vesicle isolation |
| Content And Storage | Store at 4°C. |
| Target Species | Human |
| Quantity | 10 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction