Learn More
IBA Lifesciences CD9 Fab-TACS™ Exosome Agarose Column Starter Kit
Product Code. 17207750 Shop All IBA Lifesciences Products
Click to view available options
:
Human
Product Code. 17207750
Brand: IBA Lifesciences 63319002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Label-free isolation of CD9+ exosomes from cell culture media, serum or plasma
  • Fab-TACS™ is a traceless affinity exosome isolation approach based on our Strep-tag™ technology
  • CD9 Fab-TACS™ Exosome Agarose Column Starter Kit allows label-free isolation of human CD9+ exosomes from plasma, serum and cell culture supernatants
  • CD9 is a member of the tetraspanin family present on the surface of exosomes
  • Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
  • Exosomes adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target particles are efficiently washed away
  • In a final step, the addition of biotin causes the elution of the exosomes and the Fab-Streps. After elution, the Fab-Streps self-dissociate from the exosome surface due to their low affinity
  • The label-free, fully functional exosomes are now ready for further downstream applications.
Specifications
Includes Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD9 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter
Target Species Human