missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (CD84.1.21) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £410.00
Specifications
| Antigen | CD84/SLAMF5 |
|---|---|
| Clone | CD84.1.21 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:100-1:200, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222312
|
Novus Biologicals
NB100-65929-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18226930
|
Novus Biologicals
NB100-65929 |
0.1 mg |
£410.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
| CD84/SLAMF5 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| CD84 | |
| IgG1 | |
| Protein G purified | |
| NB100-65929 recognizes the human CD84 cell surface antigen, a highly glycosylated cell surface antigen of 64-82kD. CD84 is predominantly expressed by B lymphocytes, monocytes and macrophages, although lower levels of expression have also been reported on T cells, granulocytes and platelets. |
| CD84.1.21 | |
| Flow Cytometry 1:100-1:200, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9UIB8 | |
| 8832 | |
| Mouse pre-B cell line 300.19 transfected with CD84 cDNA. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title