missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (CD84.1.21) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65929-0.025MG
This item is not returnable.
View return policy
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
| CD84/SLAMF5 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:100-1:200, Immunoprecipitation 1:10-1:500 | |
| Q9UIB8 | |
| CD84 | |
| Mouse pre-B cell line 300.19 transfected with CD84 cDNA. | |
| 0.025 mg | |
| Primary | |
| NB100-65929 recognizes the human CD84 cell surface antigen, a highly glycosylated cell surface antigen of 64-82kD. CD84 is predominantly expressed by B lymphocytes, monocytes and macrophages, although lower levels of expression have also been reported on T cells, granulocytes and platelets. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation | |
| CD84.1.21 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8832 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction