Learn More
CD83 Mouse anti-Human, APC, Clone: HB15E, BD
Mouse Monoclonal Antibody
£66.40 - £261.37
Specifications
| Antigen | CD83 |
|---|---|
| Clone | HB15E |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15851509
|
BD Biosciences
561960 |
25 Tests |
£68.50 £66.40 / Each Save £2.10 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15836508
|
BD Biosciences
551073 |
100 Tests |
£272.00 £261.37 / Each Save £10.63 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HB15e monoclonal antibody specifically binds to a 45 kDa type 1 transmembrane glycoprotein member of the Ig superfamily. CD83 is composed of a single V-type Ig extracellular domain with a C-terminal cytoplasmic tail. Cell surface CD83 is expressed mainly by follicular dendritic cells, circulating dendritic cells, interdigitating dendritic cells in lymphoid tissues, in vitro-generated dendritic cells and thymic dendritic cells. However, its expression is not restricted to dendritic cells. CD83 is also expressed on some germinal center B cells and some lymphoblastoid cell lines. Although its function is not known, it may play a role in cell-cell interaction during antigen presentation.
Specifications
| CD83 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| BL11; HB15; B-cell activation protein |
| HB15E | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human CD83 transfected COS cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.