Learn More
CD83 Mouse anti-Human, APC, Clone: HB15E, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551073
Descripción
Reacts with a 45kDa transmembrane protein member of the Ig superfamily. CD83 is composed of a single V-type Ig extracellular domain with a C-terminal cytoplasmic tail. CD83 is found mainly on follicular dendritic cells, circulating dendritic cells, interd
Host Species: Mouse
Clone: HB15e
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Especificaciones
| CD83 | |
| Monoclonal | |
| APC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CD83 transfected COS cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| HB15E | |
| BL11; HB15; B-cell activation protein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.