missing translation for 'onlineSavingsMsg'
Learn More

CD81 Mouse anti-Human, PE, Clone: JS-81, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555676

 View more versions of this product

Product Code. 15898348

  • ONLINE EXCLUSIVE PRICE
    £280.00
    £268.36 / Each
    Save £11.64
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

JS-81 identifies the newly clustered CD81 antigen, a 26kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. MAbs to CD81 have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex which can include the Leu13 molecule, and the CD19 and CD21 molecules in B cells. MAbs to CD81 are reported to induce homotypic cell aggregation.

Host Species: Mouse
Clone: JS-81
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD81
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
JS-81
TAPA-1
Mouse
100 Tests
Immunology
Canine, Human, Rabbit
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.