Learn More
CD81 Mouse anti-Human, PE, Clone: JS-81, BD
Mouse Monoclonal Antibody
£105.08 - £268.36
Specifications
| Antigen | CD81 |
|---|---|
| Clone | JS-81 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821509
|
BD Biosciences
561957 |
25 Tests |
£111.00 £105.08 / Each Save £5.92 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15898348
|
BD Biosciences
555676 |
100 Tests |
£280.00 £268.36 / Each Save £11.64 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JS-81 identifies the newly clustered CD81 antigen, a 26 kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. MAbs to CD81 have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex which can include the Leu13 molecule, and the CD19 and CD21 molecules in B cells. MAbs to CD81 are reported to induce homotypic cell aggregation.
Specifications
| CD81 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| JS-81 | |
| Monoclonal | |
| Mouse | |
| Canine, Human, Rabbit | |
| IgG1 κ | |
| Affinity Purified | |
| TAPA-1 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.