Learn More
IBA Lifesciences CD81 Fab-TACS™ Exosome Agarose Column Starter Kit
Shop All IBA Lifesciences Products
Click to view available options
:
Human
Description
- Fab-TACS™ is a traceless affinity exosome isolation approach based on our Strep-tag™ technology. CD81 Fab-TACS™ Exosome Agarose Column Starter Kit allows label-free isolation of human CD81+ exosomes from plasma, serum and cell culture supernatants. CD81 is a member of the tetraspanin family present on the surface of exosomes
- Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
- Exosomes adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target particles are efficiently washed away
- In a final step, the addition of biotin causes the elution of the exosomes and the Fab-Streps. After elution, the Fab-Streps self-dissociate from the exosome surface due to their low affinity
- The label-free, fully functional exosomes are now ready for further downstream applications
Specifications
Specifications
| Includes | Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD81 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter |
| Target Species | Human |