missing translation for 'onlineSavingsMsg'
Learn More

CD80 Armenian Hamster, FITC, Clone: 16-10A1, enQuireBio™

Armenian Hamster Monoclonal Antibody

Brand:  EnquireBio QAB50-F-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932148

  • £118.00 / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD80
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd80
Protein A/G Purified
RUO
12519
IgG
Flow Cytometry
16-10A1
FITC
XP_017172347
Armenian Hamster
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers