missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Armenian Hamster, FITC, Clone: 16-10A1, enQuireBio™
Armenian Hamster Monoclonal Antibody
£62.45 - £182.00
Specifications
Antigen | CD80 |
---|---|
Clone | 16-10A1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942148
|
EnquireBio
QAB50-F-25UG |
25 μg |
£62.45
25µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932148
|
EnquireBio
QAB50-F-100UG |
100 μg |
£118.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952148
|
EnquireBio
QAB50-F-500UG |
500 μg |
£182.00
500µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD80 | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12519 | |
IgG | |
Protein A/G Purified |
16-10A1 | |
Flow Cytometry | |
FITC | |
RUO | |
XP_017172347 | |
Cd80 | |
Primary | |
Store at 2-4°C. |