missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Baboon, Human, Rhesus, , Brilliant Ultraviolet 496, Clone: RPA-T8, BD Horizon™

Mouse Monoclonal Antibody

£241.80

Specifications

Antigen CD8
Clone RPA-T8
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 496
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17217994
View Documents
BD Biosciences
612943
25 Tests
ONLINE EXCLUSIVE PRICE
£253.00
£241.80 / 25 tests
Save £11.20
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications

Specifications

CD8
Flow Cytometry
Brilliant Ultraviolet 496
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CD8a
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
RPA-T8
Monoclonal
Mouse
Baboon, Human, Rhesus Monkey
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.