missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Baboon, Human, Rhesus, , Brilliant Ultraviolet 496, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612943
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| CD8 | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 | |
| Human CD8a | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| RPA-T8 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Baboon, Human, Rhesus Monkey | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction