missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD8 Fab-TACS™ Gravity Kit

Product Code. p-7139084 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
10 Columns, Human
4 Columns, Human
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15899784

Brand: IBA Lifesciences 63203010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IBA Lifesciences™ CD8 Fab-TACS™ Gravity Kit is for reversible cell isolation delivering label-free target cells.

  • The kit allows the selection of CD8+ T cells directly from whole blood.
  • CD8 Fab-Strep, human
  • Buffer CI (10x)
  • Biotin stock solution
  • Fab-TACS™ Gravity Adapter
  • Fab-TACS™ Flow Restrictor
  • The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
  • Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
  • Subsequently, whole blood or other blood preparations pass through the column.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
  • Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
  • After elution, the Fab-Streps self-dissociate from the cell surfaces.
  • The label-free authentic target cells are now ready for further downstream applications.

Specifications

Product Type Non-magnetic Cell Isolation Kit
Content And Storage 2°C to 8°C
Donor Source Human Whole Blood
For Use With (Application) Isolation of label-free cells
Sufficient For 500 mL whole blood
Quantity 10 Columns, Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.