missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a (B-Cell Marker) Mouse, Clone: rIGA/764, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 973-MSM11-P1
Additional Details : Weight : 0.01000kg
Specifications
CD79a (B-Cell Marker) | |
Cell surface | |
rIGA/764 | |
Unconjugated | |
Mouse | |
44kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
973 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human CD79A protein | |
Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |