missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a (B-Cell Marker) Mouse, Clone: rIGA/764, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | CD79a (B-Cell Marker) |
---|---|
Clone | rIGA/764 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941288
|
EnquireBio
973-MSM11-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931288
|
EnquireBio
973-MSM11-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD79a (B-Cell Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human CD79A protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
44kDa |
rIGA/764 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
973 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell surface | |
Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. |