missing translation for 'onlineSavingsMsg'
Learn More

CD75 Monoclonal Antibody (LN-1), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£67.25

Specifications

Antigen CD75
Clone LN-1
Concentration 5μl/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15519896
View Documents
Invitrogen
50-0759-41
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Description: This LN-1 monoclonal antibody reacts with human CD75, a cell surface carbohydrate determinant produced by alpha-2,6-sialytransferase. CD75 is expressed predominantly on mature B cells in peripheral blood and germinal centers, as well as several B cell lines (e.g., Raji, Ramos). CD75 is also expressed on non-immune tissues such as breast, prostate, colon, liver, and kidney. Reports demonstrate that CD75 is a ligand of CD22. Studies have shown that, depending on fixation and tissue processing, the LN-1 antibody also recognizes astrocytes and microglia in the human brain. Applications Reported: This LN-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This LN-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi but which can be proteolytically processed to a soluble form, is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

CD75
5μl/Test
Monoclonal
Liquid
RUO
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Sialyltransferase 1, SIAT1, ST6GalI, ST6N, B-cell antigen CD75, CMP-N-acetylneuraminate beta-galactosamide alpha-2, 6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2, 6-sialyltransferase 1, ST6Gal I, alpha 2, 6-ST 1, beta-galactoside alpha-2, 6-sialyltransferase 1, sialyltransferase 1
ST6GAL1
Primary
Store at 2-8°C. Do not freeze. Light-sensitive material.
ST6GAL1
LN-1
Flow Cytometry
eFluor 660
Mouse
Human
P15907
6480
IgM
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.