missing translation for 'onlineSavingsMsg'
Learn More

CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), Functional Grade, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

£7770.00 - £12950.00

Specifications

Antigen CD73
Clone eBioTY/11.8 (TY/11.8)
Concentration 1 mg/mL
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17111082
View Documents
Affymetrix eBioscience
16-0731-025
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17121082
View Documents
Invitrogen
16-0731-050
50 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

eBioTY/11.8 recognizes the with CD73 a 69-kDa GPI-anchored cell-surface protein with ecto-5'-nucleotidase activity. Expression on myeloid cells (CD11b) is restricted to the bone marrow. In human CD73 can be induced to secrete a soluble form with IL-2 suggesting a role in mediating activation signals. Differences between human and mouse CD73 have been reported. BALB/c mice have more CD4+CD73+ than CD8+CD73+ while the reciprocal is documented in humans. CD73 is expressed on a subset of lymphocytes and increases during lymphocyte maturation. Recently it has been found that memory CD4 T cells express and are similar to the uncommitted primed precursor helper cells (Thpp) that can differentiate into Th1 or Th2 cells. Furthermore CD73 has been found on regulatory T cells.

CD73 Monoclonal antibody specifically detects CD73 in Mouse samples. It is validated for Control, Flow Cytometry, Functional Assay
Specifications

Specifications

CD73
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
2210401F01Rik; 5 nucleotidase, ecto; 5' nucleotidase, ecto; 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; Ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; Nte; Purine 5-Prime-Nucleotidase
IgG1
Affinity chromatography
Antibody
eBioTY/11.8 (TY/11.8)
Flow Cytometry, Functional Assay
Functional Grade
Rat
Mouse
Q61503
23959
Primary
4° C
NT5E
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.