missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 / Transferrin Receptor (TFRC) (Extracellular Domain) Mouse, Clone: TFRC/1839, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7037-MSM9-P0
Additional Details : Weight : 0.01000kg
Specifications
CD71 / Transferrin Receptor (TFRC) (Extracellular Domain) | |
Cell surface | |
TFRC/1839 | |
Unconjugated | |
Mouse | |
85-95kDa (monomer); 190kDa (dimer) | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7037 | |
IgG2b κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human extracellular TFRC protein fragment (apprx. aa 94-212) | |
Jurkat, HeLa, MCF-7 or K562 cells. Human placenta or Bone Marrow. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |