missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 / Transferrin Receptor (TFRC) (Extracellular Domain) Mouse, Clone: TFRC/1839, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | CD71 / Transferrin Receptor (TFRC) (Extracellular Domain) |
---|---|
Clone | TFRC/1839 |
Concentration | 200μg/mL |
Applications | ELISA, Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940948
|
EnquireBio
7037-MSM9-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930948
|
EnquireBio
7037-MSM9-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD71 / Transferrin Receptor (TFRC) (Extracellular Domain) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human extracellular TFRC protein fragment (apprx. aa 94-212) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
85-95kDa (monomer); 190kDa (dimer) |
TFRC/1839 | |
ELISA, Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
7037 | |
IgG2b κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell surface | |
Jurkat, HeLa, MCF-7 or K562 cells. Human placenta or Bone Marrow. |