missing translation for 'onlineSavingsMsg'
Learn More

CD71 Mouse anti-Human, APC, Clone: M-A712, BD

Mouse Monoclonal Antibody

£111.86 - £213.74

Specifications

Antigen CD71
Clone M-A712
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15851489
View Documents
BD Biosciences
561940
25 Tests
ONLINE EXCLUSIVE PRICE
£117.00
£111.86 / Each
Save £5.14
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15816628
View Documents
BD Biosciences
551374
100 Tests
ONLINE EXCLUSIVE PRICE
£223.00
£213.74 / Each
Save £9.26
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The M-A712 monoclonal antibody specifically binds to CD71 which is also known as the transferrin receptor (TFR). This type II transmembrane glycoprotein is expressed on cells as a disulfide-linked homodimer comprised of 95 kDa monomers. CD71 is expressed on activated lymphocytes, monocytes, macrophages, erythroid progenitors, brain endothelium, and most proliferating cells. CD71 is not expressed on resting lymphocytes and is upregulated during lymphocyte responses to antigens or mitogens. Through an endocytic pathway, the transferrin receptor mediates cellular iron uptake by binding and internalizing iron that is bound to transferrin. After releasing iron within the low pH endosomal environment, transferrin and its receptor can be recycled to the cell surface.

Specifications

Specifications

CD71
Flow Cytometry
APC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
TFR; TFRC; Trfr; TfR1; Transferrin receptor protein 1
M-A712
Monoclonal
Mouse
Human
T-CLL and Jurkat Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.