missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Mouse anti-Human, Clone: T3-3A1, Novus Biologicals™
Mouse Monoclonal Antibody
£448.00
Specifications
| Antigen | CD7 |
|---|---|
| Clone | T3-3A1 |
| Dilution | Flow Cytometry 0.5 - 1ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
Description
CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, CyTOF-ready.Specifications
| CD7 | |
| Flow Cytometry 0.5 - 1ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| CD7 | |
| IgG1 κ | |
| Protein A or G purified | |
| Recognizes a protein of 40kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. |
| T3-3A1 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Signal Transduction | |
| 10mM PBS with No Preservative | |
| 924 | |
| Human T cells | |
| Primary | |
| Store at 4°C. | |
| 40 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title