missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Mouse anti-Human, Clone: T3-3A1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54901-0.1MG
This item is not returnable.
View return policy
Description
CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, CyTOF-ready.
Specifications
| CD7 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS with No Preservative | |
| CD7 | |
| Human T cells | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 40kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| T3-3A1 | |
| Flow Cytometry 0.5 - 1ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| Mouse | |
| 40 kDa | |
| 0.1 mg | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction