Learn More
CD69 Hamster anti-Mouse, PE, Clone: H1.2F3, BD
Hamster Monoclonal Antibody
£88.51 - £399.24
Specifications
| Applications | Flow Cytometry |
|---|---|
| Classification | Monoclonal |
| Regulatory Status | RUO |
| Target Species | Mouse |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15871479
|
BD Biosciences
561932 |
25 μg |
£92.05 £88.51 / Each Save £3.54 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15861489
|
BD Biosciences
561941 |
0.1 mg |
£148.00 £142.18 / Each Save £5.82 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15878058
|
BD Biosciences
555318 |
0.5 mg |
£416.00 £399.24 / Each Save £16.76 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The In-1 monoclonal antibody specifically binds to CD74, the MHC class II-associated Invariant chain (Ii), expressed in I-A+ cell populations. Although it is predominantly expressed intracellularly, it has also been detected on the cell surface of some B-cell lines and at low levels on a small number of B lymphocytes. In the mouse, alternatively spliced transcripts encode 31-kDa and 41-kDa isoforms, and the cell-surface CD74 has a chondroitin sulfate side chain which allows it to interact with CD44. CD74 is directly involved in the intracellular transport of MHC class II molecules and antigen presentation, and it affects the maturation of T and B lymphocytes.
Specifications
| Flow Cytometry | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified |
| Monoclonal | |
| Mouse | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.