missing translation for 'onlineSavingsMsg'
Learn More

CD69 Hamster anti-Mouse, PE, Clone: H1.2F3, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 561932

 View more versions of this product

Product Code. 15871479

  • ONLINE EXCLUSIVE PRICE
    £92.05
    £88.51 / Each
    Save £3.54
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The H1.2F3 monoclonal antibody specifically binds to CD69 (Very Early Activation antigen), an 85 kDa disulfide-linked homodimer of differentially glycosylated subunits. CD69 is a C-type lectin, most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. Its expression is rapidly induced upon activation of lymphocytes (T, B, NK, and NK-T cells), neutrophils, and macrophages. CD69 is expressed also on thymocytes that are undergoing positive selection; its role in that process is unclear. H1.2F3 mAb augments PMA-induced T-cell stimulation and IFN-γ-induced macrophage stimulation. IL-2-activated NK cells express CD69, and H1.2F3 mAb induces redirected lysis of FcR-bearing target cells by NK cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD69
Monoclonal
0.2mg/mL
VEA; Very Early Activation Antigen; AIM; Activation Induced Molecule
Hamster
Affinity Purified
RUO
Mouse
IgG1 λ3
Flow Cytometry
H1.2F3
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Dendritic Epidermal T Cell Line Y245
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.