missing translation for 'onlineSavingsMsg'
Learn More

CD68 Rat anti-Mouse, PE, Clone: FA/11 (also known as FA-11, FA11), BD Pharmingen™

Rat Monoclonal Antibody

£119.58 - £280.34

Specifications

Antigen CD68
Clone FA/11 (also known as FA-11, FA11)
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15835009
View Documents
BD Biosciences
566387
25 μg
ONLINE EXCLUSIVE PRICE
£126.00
£119.58 / Each
Save £6.42
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825009
View Documents
BD Biosciences
566386
0.1 mg
ONLINE EXCLUSIVE PRICE
£284.00
£280.34 / Each
Save £3.66
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD68 Monoclonal antibody specifically detects CD68 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD68
0.2 mg/mL
Monoclonal
Purified
RUO
IgG2a κ
Refrigerate (2 to 8°C)
FA/11 (also known as FA-11, FA11)
Flow Cytometry
PE
Rat
Mouse
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.