missing translation for 'onlineSavingsMsg'
Learn More

CD68 Rat anti-Mouse, PE, Clone: FA/11 (also known as FA-11, FA11), BD Pharmingen™

Rat Monoclonal Antibody

Brand:  BD Biosciences 566387

 View more versions of this product

Product Code. 15835009

  • ONLINE EXCLUSIVE PRICE
    £126.00
    £119.58 / Each
    Save £6.42
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD68 Monoclonal antibody specifically detects CD68 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD68
Monoclonal
0.2 mg/mL
Rat
RUO
Mouse
Purified
Flow Cytometry
FA/11 (also known as FA-11, FA11)
PE
25 μg
Primary
Refrigerate (2 to 8°C)
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.