Learn More
Invitrogen™ CD68 Monoclonal Antibody (eBioY1/82A (Y1/82A)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£1376.00 - £3115.00
Specifications
| Antigen | CD68 |
|---|---|
| Clone | eBioY1/82A (Y1/82A) |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
The eBioY1/82 antibody reacts with CD68 which belongs to the sialomucin family and is closely related to the family of lysosomal-associated membrane proteins (lamps) and scavenger receptor. CD68 is predominantly an intracellular protein, found mainly in the late endosomal compartment but can be detected in smaller amounts on the surface of cells of mainly myeloid derived cells; monocytes and macrophages, dendritic cells (and in some langerhan cells), neutrophils, basophils mast cells, myeloid progenitor cells, and a subset of CD34+ hemopoietic bone marrow progenitor cells. The function has not been fully elucidated but based on it's homolgy and structure suggests a role in antigen processing or presentation and protection of the lysosomal membrane from hydrolytic enzymes. Reports have also seen expression in activated T cells, and about 40% of peripheral blood B-lymphocytes and 50% of all B-ALL. Staining with eBioY1/82A shows surface staining on B cells and a minor percentage of monocytes while intracellular staining results in about 50% of B cells and most monocytes.
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Specifications
| CD68 | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| P34810 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| eBioY1/82A (Y1/82A) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 968 | |
| Primary | |
| 4°C | |
| Cd68 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.