missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD66 Monoclonal Antibody (CD66a-B1.1), Super Bright™ 702, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£142.00 - £354.00
Specifications
| Antigen | CD66 |
|---|---|
| Clone | CD66a-B1.1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16351634
|
Affymetrix eBioscience
67-0668-41 |
25 Tests | |||||||
16361634
![]() |
Invitrogen™
67-0668-42 |
100 Tests |
£354.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
The CD66 molecules are 180-200 kDa glycoproteins of carcinoembryonic antigen family. They are present on all blood granulocytes and some tissue macrophages, but are absent from other hematopoietic cells. The expression of CD66 increases significantly on granulocytes upon their activation. CD66a (BGP1, CEACAM1) and CD66d (CGM1, CEACAM3), as well as CEACAM2 and 4, are transmembrane proteins, whereas CD66c (CEAL, NCA, CEACAM6) and CD66e (CEA, CEACAM5), as well as CEACAM7 and 8, are anchored to the plasma membrane by C-terminal glycosylphosphatidylinositol (GPI) lipid moiety.Specifications
| CD66 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 1048, 1084, 1089, 4680, 634 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| CD66a-B1.1 | |
| Flow Cytometry | |
| Super Bright 702 | |
| Mouse | |
| Human | |
| O75871, P06731, P13688, P40198, P40199 | |
| CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title