missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD66 Monoclonal Antibody (CD66a-B1.1), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 67-0668-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD66 Monoclonal antibody specifically detects CD66 in Human samples. It is validated for Flow Cytometry
Specifications
| CD66 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| O75871, P06731, P13688, P40198, P40199 | |
| CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6 | |
| Affinity chromatography | |
| RUO | |
| 1048, 1084, 1089, 4680, 634 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| CD66a-B1.1 | |
| Super Bright 702 | |
| CEACAM3 | |
| BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1; Carcinoembryonic antigen CGM6; Carcinoembryonic antigen CGM7; Carcinoembryonic antigen-related cell adhesion molecule 1; Carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 4; Carcinoembryonic antigen-related cell adhesion molecule 5; Carcinoembryonic antigen-related cell adhesion molecule 6; Carcinoembryonic antigen-related cell adhesion molecule 8; CD66a; CD66b; CD66c; CD66D; CD66e; CD67; CD67 antigen; CEA; CEACAM1; CEACAM-1; CEACAM3; CEACAM-3; CEACAM4; CEACAM5; CEACAM-5; CEACAM6; CEACAM-6; CEACAM8; CEAL; CGM1; CGM6; CGM7; CGM7_HUMAN; Meconium antigen 100; NCA; NCA-95; Non-specific crossreacting antigen; Non-specific cross-reacting antigen NCA-95; Non-specific cross-reacting antigen W236; Normal cross-reacting antigen; pan CEACAM; W264; W282 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction