missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD6 Recombinant Rabbit Monoclonal Antibody (021), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£113.00 - £359.00
Specifications
| Antigen | CD6 |
|---|---|
| Clone | 021 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019561
![]() |
Invitrogen™
MA552415 |
100 Tests |
£359.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019586
|
Invitrogen™
MA552416 |
25 Tests |
£113.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD6 Recombinant Monoclonal antibody specifically detects CD6 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD6 | |
| 0.1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.09% sodium azide | |
| Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120 | |
| Recombinant Mouse CD6 protein (Gly 17-Gly396). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD6 |
| 021 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Mouse | |
| Q61003 | |
| 12511 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title