missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD59 Mouse anti-Human, Clone: MEM43, Novus Biologicals™
Mouse Monoclonal Antibody
£405.00 - £822.00
Specifications
| Antigen | CD59 |
|---|---|
| Clone | MEM43 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18322057
|
Novus Biologicals
NBP3-14606-200UL |
200 μg |
£405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18370874
|
Novus Biologicals
NBP3-14606-500UL |
500 μg |
£822.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
| CD59 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| PBS. Sterile-filtered through 0.22 um. | |
| 966 | |
| IgG2a κ | |
| Affinity purified | |
| Antibody |
| MEM43 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein | |
| Thymocytes and T lymphocytes | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title