missing translation for 'onlineSavingsMsg'
Learn More

CD59 Mouse anti-Human, Clone: MEM43, Novus Biologicals™

Product Code. 18370874 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370874 500 μg 500µL
18322057 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370874 Supplier Novus Biologicals Supplier No. NBP314606500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD59
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MEM43
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Host Species Mouse
Immunogen Thymocytes and T lymphocytes
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.