missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD59 Antibody (Quark patent anti-CD59), Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen CD59
Clone Quark patent anti-CD59
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30048426
View Documents
Novus Biologicals™
NBP3-28046-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30047170
View Documents
Novus Biologicals™
NBP3-28046-100ug
100 μg
£257.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046216
View Documents
Novus Biologicals™
NBP3-28046-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

CD59
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Adaptive Immunity, Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
CD59
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Quark patent anti-CD59
Monoclonal
Purified
RUO
Human
P13987
966
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.