missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD59 Antibody (Quark patent anti-CD59), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28046-50ug
This item is not returnable.
View return policy
Description
CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD59 | |
| Monoclonal | |
| Unconjugated | |
| P13987 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Quark patent anti-CD59 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein | |
| CD59 | |
| 50 μg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 966 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction