missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (IHC539), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75981-1ml
This item is not returnable.
View return policy
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD57 | |
| Monoclonal | |
| Unconjugated | |
| Tris Buffer, pH 7.3 - 7.7, with 1% BSA | |
| Mouse | |
| Protein L purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| IHC539 | |
| Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant Human CD57 | |
| 1 mL | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction