missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (IHC539), Novus Biologicals™
Mouse Monoclonal Antibody
£143.00 - £373.00
Specifications
| Antigen | CD57 |
|---|---|
| Clone | IHC539 |
| Dilution | Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692941
|
Novus Biologicals
NBP2-75981-1ml |
1 mL |
£373.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643721
|
Novus Biologicals
NBP2-75981-0.1ml |
0.1 mL |
£143.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD57 | |
| Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant Human CD57 | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. |
| IHC539 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Immunology | |
| Tris Buffer, pH 7.3 - 7.7, with 1% BSA | |
| 27087 | |
| IgM κ | |
| Protein L purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title