Learn More
Invitrogen™ CD56 (NCAM) Recombinant Mouse Monoclonal Antibody (CMSSB), Alexa Fluor™ Plus 488, Invitrogen™
Mouse Recombinant Monoclonal Antibody
£188.00 - £360.00
Specifications
| Antigen | CD56 (NCAM) |
|---|---|
| Clone | CMSSB |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30131983
|
Invitrogen™
740024MP48820UG |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30131622
|
Invitrogen™
740024MP488 |
50 μL |
£360.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This CMSSB antibody has been tested by IHC and flow cytometric analysis of NK-92cells. This may be used for IHC at 10 μg/mL and for FC at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 494 nm; Emission: 519 nm; Laser: Blue Laser Filtration: 0.2 μm post-manufacturing filtered.
CD56 (NCAM) Recombinant Monoclonal antibody specifically detects CD56 (NCAM) in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| CD56 (NCAM) | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| CMSSB | |
| Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor Plus 488 | |
| Mouse | |
| Human | |
| P13591 | |
| 4684 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ncam1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.