missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Mouse anti-Human , BUV615, Clone: NCAM16.2 (also known as NCAM 16), BD Biosciences™
Mouse Monoclonal Antibody
£224.85 - £445.74
Specifications
| Antigen | CD56 |
|---|---|
| Clone | NCAM16.2 (also known as NCAM 16) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 615 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17228044
|
BD Biosciences
613002 |
25 Tests |
£235.00 £224.85 / 25 tests Save £10.15 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17218044
|
BD Biosciences
613001 |
100 Tests |
£451.00 £445.74 / 100 tests Save £5.26 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow CytometrySpecifications
| CD56 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 615 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Immunoaffinity-enriched adult human brain NCAM | |
| Primary |
| NCAM16.2 (also known as NCAM 16) | |
| Monoclonal | |
| Mouse | |
| Human | |
| NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 | |
| IgG2b κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title