missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Mouse anti-Human , BUV615, Clone: NCAM16.2 (also known as NCAM 16), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 613001
This item is not returnable.
View return policy
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry
Specifications
| CD56 | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 | |
| Immunoaffinity-enriched adult human brain NCAM | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| NCAM16.2 (also known as NCAM 16) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction