Learn More
CD55 Mouse anti-Human, FITC, Clone: IA10, BD
Mouse Monoclonal Antibody
£567.89 - £1425.00
Specifications
| Antigen | CD55 |
|---|---|
| Clone | IA10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15851449
|
BD Biosciences
561900 |
25 Tests | |||||||
|
15848368
|
BD Biosciences
555693 |
100 Tests |
£595.00 £567.89 / Each Save £27.11 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with CD55, decay-accelerating factor (DAF), a glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein of approximately 70 kDa, expressed on hematopoietic cells. It has been suggested that the role of DAF is to protect cells from damage by autologous complement preventingthe amplification steps of the complement cascade by interfering with the assembly of the C3-convertases, C4b2a and C3bBb, and the C5-convertases, C4b2a3b and C3bBb3b.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD55 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| IA10 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2a κ | |
| Affinity Purified | |
| DAF |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.