Learn More
CD55 Mouse anti-Human, FITC, Clone: IA10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561900
Description
Reacts with CD55, decay-accelerating factor (DAF), a glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein of approximately 70 kDa, expressed on hematopoietic cells. It has been suggested that the role of DAF is to protect cells from damage by autologous complement preventingthe amplification steps of the complement cascade by interfering with the assembly of the C3-convertases, C4b2a and C3bBb, and the C5-convertases, C4b2a3b and C3bBb3b.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD55 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| IA10 | |
| DAF | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.