missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD54 / ICAM-1 Mouse, PE, Clone: 15.2, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB47-PE-25Tests
Additional Details : Weight : 0.01000kg
Specifications
CD54 / ICAM-1 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
ICAM1 | |
Protein A/G Purified | |
RUO | |
3383 | |
IgG1 |
Flow Cytometry | |
15.2 | |
PE | |
NP_000192 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |