missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD54 / ICAM-1 Mouse, PE, Clone: 15.2, enQuireBio™
Mouse Monoclonal Antibody
£99.15 - £237.00
Specifications
Antigen | CD54 / ICAM-1 |
---|---|
Clone | 15.2 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952098
|
EnquireBio
QAB47-PE-25TESTS |
25 Tests |
£99.15
25 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942098
|
EnquireBio
QAB47-PE-100TESTS |
100 Tests |
£237.00
100 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD54 / ICAM-1 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3383 | |
IgG1 | |
Protein A/G Purified |
15.2 | |
Flow Cytometry | |
PE | |
RUO | |
NP_000192 | |
ICAM1 | |
Primary | |
Store at 2-4°C. |