missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (A25-G), Novus Biologicals™
Rabbit Monoclonal Antibody
£442.00
Specifications
| Antigen | CD5 |
|---|---|
| Clone | A25-G |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD5 | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200 | |
| Monoclonal | |
| Rabbit | |
| Human | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| CD5 | |
| Primary | |
| Store at 4C. Do not freeze. |
| A25-G | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| 921 | |
| Peptide derived from the C-terminal sequence of human CD10. | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title