missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (A25-G), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-78994-100UL
This item is not returnable.
View return policy
Description
CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD5 | |
| Monoclonal | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| CD5 | |
| Peptide derived from the C-terminal sequence of human CD10. | |
| 100 μL | |
| Primary | |
| Human |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| A25-G | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200 | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 921 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction