missing translation for 'onlineSavingsMsg'
Learn More

CD47 Rat anti-Mouse, Clone: miap301, Novus Biologicals™

Rat Monoclonal Antibody

£244.00 - £333.00

Specifications

Antigen CD47
Clone miap301
Concentration 0.5 mg/ml
Dilution Western Blot, Flow Cytometry 0.5 μg/106 cells, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen
Applications Western Blot
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18572274
View Documents
Novus Biologicals
NBP1-43407-0.5mg
0.5 mg
£333.00
0.50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18553472
View Documents
Novus Biologicals
NBP1-43407-0.1mg
0.1 mg
£244.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18546661
View Documents
Novus Biologicals
NBP1-43407-0.05mg
0.05 mg
£246.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD47 Monoclonal specifically detects CD47 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

CD47
0.5 mg/ml
Western Blot
Unconjugated
Rat
Immunology, Signal Transduction
Aqueous, may contain carrier protein/stabilizer.
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
The immunogen for this antibody was CD47.
Primary
Store at 4°C. Do not freeze.
miap301
Western Blot, Flow Cytometry 0.5 μg/106 cells, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen
Monoclonal
Purified
RUO
Mouse
Q08722
961
IgG2a κ
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.