missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Rat anti-Mouse, Clone: miap301, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43407-0.1mg
This item is not returnable.
View return policy
Description
CD47 Monoclonal specifically detects CD47 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| CD47 | |
| Monoclonal | |
| 0.5 mg/ml | |
| Western Blot, Flow Cytometry 0.5 μg/106 cells, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen | |
| Q08722 | |
| Rat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| miap301 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| The immunogen for this antibody was CD47. | |
| 0.1 mg | |
| Immunology, Signal Transduction | |
| 961 | |
| Store at 4°C. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction